missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl 4-Hydroxyisophthalate 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 97°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 210.19 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dimethyl 4-Hydroxyisophthalate |
Safety and Handling
| TSCA | Yes |