missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 4-Hydroxyisophthalate 97.0+%, TCI America™

Catalog No. D35621G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35621G 1 g
D35625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35621G Supplier TCI America Supplier No. D35621G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5985-24-0
Molecular Formula C10H10O5
Molecular Weight (g/mol) 210.185
MDL Number MFCD00152444
InChI Key ALBUJVBOIXVVLS-UHFFFAOYSA-N
Synonym 4-Hydroxyisophthalic Acid Dimethyl Ester
PubChem CID 80087
IUPAC Name dimethyl 4-hydroxybenzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=C(C=C1)O)C(=O)OC

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 1 g
Formula Weight 210.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 4-Hydroxyisophthalate