missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride, 97%

Catalog No. AAB2002803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2002803 1 g
AAB2002806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2002803 Supplier Thermo Scientific Chemicals Supplier No. B2002803

CAS: 121071-71-4 | C8H10ClNO4S | 251.681 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121071-71-4
Molecular Formula C8H10ClNO4S
Molecular Weight (g/mol) 251.681
MDL Number MFCD00205189
InChI Key RETIFGMAGNDLAN-UHFFFAOYSA-N
PubChem CID 2808333
IUPAC Name dimethyl 4-aminothiophene-2,3-dicarboxylate;hydrochloride
SMILES COC(=O)C1=C(SC=C1N)C(=O)OC.Cl

Specifications

Melting Point 166°C to 169°C
Quantity 1 g
Sensitivity Hygroscopic
Synonym dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride, 2,3-dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride, acmc-209v6b, dimethyl 4-aminothiophene-2,3-dicarboxylate hcl, dimethyl 4-aminothiophene-2,3-dicarboxylate hydroc, 3-amino-4,5-bismethoxycarbonylthiophene hydrochloride, dimethyl4-aminothiophene-2,3-dicarboxylate hydrochloride, 4-aminothiophene-2,3-dicarboxylic acid dimethyl ester hydrochloride, methyl 4-amino-2-methoxycarbonyl thiophene-3-carboxylate, chloride
Formula Weight 251.69
Percent Purity 97%
Chemical Name or Material Dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only