accessibility menu, dialog, popup

UserName

CAS RN 121071-71-4

IUPAC Name: 2,3-dimethyl 4-aminothiophene-2,3-dicarboxylate
Synonyms: 2,3-dimethyl 4-aminothiophene-2,3-dicarboxylate
Molecular Weight (g/mol): 215.22
Molecular Formula: C8H9NO4S
InChi Key: XQPDCUCAKYNVAO-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C(=O)OC)C(N)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results