missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl 3-Nitrophthalate 99.0+%, TCI America™
$298.25 - $870.51
Chemical Identifiers
| CAS | 13365-26-9 |
|---|---|
| Molecular Formula | C10H9NO6 |
| Molecular Weight (g/mol) | 239.183 |
| MDL Number | MFCD00017184 |
| InChI Key | MLQMIKSBTAZNBK-UHFFFAOYSA-N |
| PubChem CID | 249912 |
| IUPAC Name | dimethyl 3-nitrobenzene-1,2-dicarboxylate |
| SMILES | COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])C(=O)OC |
Chemical Identifiers
| 13365-26-9 | |
| 239.183 | |
| MLQMIKSBTAZNBK-UHFFFAOYSA-N | |
| dimethyl 3-nitrobenzene-1,2-dicarboxylate |
| C10H9NO6 | |
| MFCD00017184 | |
| 249912 | |
| COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])C(=O)OC |
Specifications
| 13365-26-9 | |
| White-Yellow | |
| MFCD00017184 | |
| dimethyl 3-nitrophthalate, 3-nitrophthalic acid dimethyl ester, 3-nitro-phthalic acid dimethyl ester, 1,2-benzenedicarboxylic acid, 3-nitro-, 1,2-dimethyl ester, 1,2-dimethyl 3-nitrophthalate, dimethyl nitrophthalate, dimethyl3-nitrophthalate, maybridge1_000481, acmc-20al75, methyl 2-methoxycarbonyl-3-nitrobenzoate | |
| COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])C(=O)OC | |
| 239.183 | |
| 239.18 | |
| Crystalline Powder |
| 68°C | |
| C10H9NO6 | |
| 5 g | |
| MLQMIKSBTAZNBK-UHFFFAOYSA-N | |
| dimethyl 3-nitrobenzene-1,2-dicarboxylate | |
| 249912 | |
| ≥99.0% (GC) | |
| Dimethyl 3-Nitrophthalate |
Safety and Handling
TSCA : No