missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 3-Nitrophthalate 99.0+%, TCI America™

Catalog No. D53105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53105G 5 g
D531025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53105G Supplier TCI America Supplier No. D53105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13365-26-9
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.183
MDL Number MFCD00017184
InChI Key MLQMIKSBTAZNBK-UHFFFAOYSA-N
PubChem CID 249912
IUPAC Name dimethyl 3-nitrobenzene-1,2-dicarboxylate
SMILES COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])C(=O)OC

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 5 g
Synonym dimethyl 3-nitrophthalate, 3-nitrophthalic acid dimethyl ester, 3-nitro-phthalic acid dimethyl ester, 1,2-benzenedicarboxylic acid, 3-nitro-, 1,2-dimethyl ester, 1,2-dimethyl 3-nitrophthalate, dimethyl nitrophthalate, dimethyl3-nitrophthalate, maybridge1_000481, acmc-20al75, methyl 2-methoxycarbonyl-3-nitrobenzoate
Formula Weight 239.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 3-Nitrophthalate