missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 3-nitrophthalate, 98%

Catalog No. AAL0671514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0671514 25 g
AAL0671506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0671514 Supplier Thermo Scientific Chemicals Supplier No. L0671514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13365-26-9 | C10H9NO6 | 239.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13365-26-9
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.183
MDL Number MFCD00017184
InChI Key MLQMIKSBTAZNBK-UHFFFAOYSA-N
PubChem CID 249912
IUPAC Name dimethyl 3-nitrobenzene-1,2-dicarboxylate
SMILES COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])C(=O)OC

Specifications

Melting Point 67°C to 70°C
Quantity 25 g
Beilstein 1885570
Synonym dimethyl 3-nitrophthalate, 3-nitrophthalic acid dimethyl ester, 3-nitro-phthalic acid dimethyl ester, 1,2-benzenedicarboxylic acid, 3-nitro-, 1,2-dimethyl ester, 1,2-dimethyl 3-nitrophthalate, dimethyl nitrophthalate, dimethyl3-nitrophthalate, maybridge1_000481, acmc-20al75, methyl 2-methoxycarbonyl-3-nitrobenzoate
Formula Weight 239.19
Percent Purity 98%
Chemical Name or Material Dimethyl 3-nitrophthalate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only