missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (2R,3R)-2,3-O-(1-Phenylethylidene)-L-tartrate, TCI America™

Catalog No. P11895G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11895G Supplier TCI America Supplier No. P11895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 104333-83-7
Molecular Formula C14H16O6
Molecular Weight (g/mol) 280.276
MDL Number MFCD00191680
InChI Key XFSCUQFGXSPZCK-GHMZBOCLSA-N
Synonym Dimethyl (4R,5R)-2-Methyl-2-phenyl-1,3-dioxolane-4,5-dicarboxylate, (2R,3R)-2,3-O-(1-Phenylethylidene)-L-tartaric Acid Dimethyl Ester
PubChem CID 395121
IUPAC Name dimethyl (4R,5R)-2-methyl-2-phenyl-1,3-dioxolane-4,5-dicarboxylate
SMILES CC1(OC(C(O1)C(=O)OC)C(=O)OC)C2=CC=CC=C2

Specifications

Quantity 5 g
Formula Weight 280.28
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl (2R,3R)-2,3-O-(1-Phenylethylidene)-L-tartrate