CAS RN 104333-83-7
IUPAC Name:
4,5-dimethyl 2-methyl-2-phenyl-1,3-dioxolane-4,5-dicarboxylate
Synonyms:
4,5-dimethyl 2-methyl-2-phenyl-1,3-dioxolane-4,5-dicarboxylate
Molecular Weight (g/mol):
280.28
Molecular Formula:
C14H16O6
InChi Key:
XFSCUQFGXSPZCK-UHFFFAOYNA-N
SMILES:
COC(=O)C1OC(C)(OC1C(=O)OC)C1=CC=CC=C1
1
–
1
of
1
results
1
Dimethyl (2R,3R)-2,3-O-(1-Phenylethylidene)-L-tartrate, TCI America™
CAS: 104333-83-7 Molecular Formula: C14H16O6 Molecular Weight (g/mol): 280.276 MDL Number: MFCD00191680 InChI Key: XFSCUQFGXSPZCK-GHMZBOCLSA-N Synonym: Dimethyl (4R,5R)-2-Methyl-2-phenyl-1,3-dioxolane-4,5-dicarboxylate, (2R,3R)-2,3-O-(1-Phenylethylidene)-L-tartaric Acid Dimethyl Ester PubChem CID: 395121 IUPAC Name: dimethyl (4R,5R)-2-methyl-2-phenyl-1,3-dioxolane-4,5-dicarboxylate SMILES: CC1(OC(C(O1)C(=O)OC)C(=O)OC)C2=CC=CC=C2