missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl (2-oxopropyl)phosphonate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 177610250
| Quantity | 25g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for C-C Bond Formation (Olefination)Chemical Identifiers
| C5H11O4P | |
| MFCD00008769 | |
| dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone | |
| 1-dimethoxyphosphorylpropan-2-one |
Specifications
| Dimethyl (2-oxopropyl)phosphonate | |
| 76°C to 79°C (3.0mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4380 to 1.441 | |
| 25g | |
| dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone | |
| CC(=O)CP(=O)(OC)OC | |
| 166.11 | |
| 166.11 |
| 4202-14-6 | |
| 93°C | |
| 95% | |
| C5H11O4P | |
| CH3COCH2P(O)(OCH3)2 | |
| MFCD00008769 | |
| UOWIYNWMROWVDG-UHFFFAOYSA-N | |
| 1-dimethoxyphosphorylpropan-2-one | |
| 77872 | |
| 95% |
Safety and Handling
EINECSNumber : 224-110-5
Promotions
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.