missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (2-oxopropyl)phosphonate, 95%, Thermo Scientific™

Catalog No. p-3736332
Change view
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC177610050 5g Glass bottle
AC177610250 25g Glass bottle
AC177611000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC177610050 Supplier Thermo Scientific Chemicals Supplier No. 177610050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 4202-14-6
Molecular Formula C5H11O4P
Molecular Weight (g/mol) 166.11
MDL Number MFCD00008769
InChI Key UOWIYNWMROWVDG-UHFFFAOYSA-N
Synonym dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone
PubChem CID 77872
IUPAC Name 1-dimethoxyphosphorylpropan-2-one
SMILES CC(=O)CP(=O)(OC)OC

Specifications

Boiling Point 76°C to 79°C (3.0mmHg)
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.4380 to 1.441
Linear Formula CH3COCH2P(O)(OCH3)2
Quantity 5g
Formula Weight 166.11
Percent Purity 95%
Chemical Name or Material Dimethyl (2-oxopropyl)phosphonate
Show More Show Less
EINECSNumber 224-110-5

RUO – Research Use Only