missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (2-Hydroxyethyl)phosphonate, TCI America™

Catalog No. p-7130719
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H07795G Supplier TCI America Supplier No. H07795G
Only null left

Chemical Identifiers

CAS 54731-72-5
Molecular Formula C4H11O4P
Molecular Weight (g/mol) 154.10
MDL Number MFCD00043376
InChI Key TZPPDWDHNIMTDQ-UHFFFAOYSA-N
Synonym dimethyl 2-hydroxyethylphosphonate, dimethyl 2-hydroxyethyl phosphonate, 2-hydroxyethylphosphonic acid dimethyl ester, 2-hydroxyethyl phosphonic acid dimethyl ester, acmc-1ao1t, 2-diethylphosphono ethan-1-ol, tzppdwdhnimtdq-uhfffaoysa, hydroxyethyldimethylphosphonic acid, 2-dimethoxyphosphoryl ethan-1-ol
PubChem CID 2733833
IUPAC Name dimethyl (2-hydroxyethyl)phosphonate
SMILES COP(=O)(CCO)OC

Specifications

Color Colorless
Boiling Point 97°C
Quantity 5 g
Formula Weight 154.10
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl (2-Hydroxyethyl)phosphonate