accessibility menu, dialog, popup

UserName

CH3OPOHOOH3C

CAS RN 54731-72-5

IUPAC Name: dimethyl (2-hydroxyethyl)phosphonate
Synonyms: dimethyl 2-hydroxyethylphosphonate
Molecular Weight (g/mol): 154.1
Molecular Formula: C4H11O4P
InChi Key: TZPPDWDHNIMTDQ-UHFFFAOYSA-N
SMILES: COP(=O)(CCO)OC
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)