missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl 2,6-Pyridinedicarboxylate 98.0+%, TCI America™
Supplier: TCI America P144425G
Specifications
| Dimethyl 2,6-Pyridinedicarboxylate | |
| 124°C | |
| C9H9NO4 | |
| 25 g | |
| SNQQJEJPJMXYTR-UHFFFAOYSA-N | |
| dimethyl pyridine-2,6-dicarboxylate | |
| 79549 | |
| ≥98.0% (GC) |
| 5453-67-8 | |
| White-Yellow | |
| MFCD00134493 | |
| dimethyl 2,6-pyridinedicarboxylate, 2,6-dimethyl pyridine-2,6-dicarboxylate, pyridine-2,6-dicarboxylic acid dimethyl ester, 2,6-pyridinedicarboxylic acid dimethyl ester, 2,6-pyridinedicarboxylic acid, dimethyl ester, dimethyl dipicolinate, dimethyl pyridine-2,6-carboxylate, chembl72405, dimethylpyridine-2,6-dicarboxylate, dipicolinic acid dimethyl ester | |
| COC(=O)C1=NC(=CC=C1)C(=O)OC | |
| 195.174 | |
| 195.17 | |
| Crystalline Powder |
Chemical Identifiers
| 5453-67-8 | |
| 195.174 | |
| SNQQJEJPJMXYTR-UHFFFAOYSA-N | |
| 79549 | |
| COC(=O)C1=NC(=CC=C1)C(=O)OC |
| C9H9NO4 | |
| MFCD00134493 | |
| dimethyl 2,6-pyridinedicarboxylate, 2,6-dimethyl pyridine-2,6-dicarboxylate, pyridine-2,6-dicarboxylic acid dimethyl ester, 2,6-pyridinedicarboxylic acid dimethyl ester, 2,6-pyridinedicarboxylic acid, dimethyl ester, dimethyl dipicolinate, dimethyl pyridine-2,6-carboxylate, chembl72405, dimethylpyridine-2,6-dicarboxylate, dipicolinic acid dimethyl ester | |
| dimethyl pyridine-2,6-dicarboxylate |
Safety and Handling
TSCA : No