missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2,6-Pyridinedicarboxylate 98.0+%, TCI America™

Catalog No. p-7135140
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
P14445G 5 g
P144425G 25 g
2 options

Catalog No. P14445G

Supplier: TCI America P14445G

Only null left

Chemical Identifiers

CAS 5453-67-8
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00134493
InChI Key SNQQJEJPJMXYTR-UHFFFAOYSA-N
Synonym dimethyl 2,6-pyridinedicarboxylate, 2,6-dimethyl pyridine-2,6-dicarboxylate, pyridine-2,6-dicarboxylic acid dimethyl ester, 2,6-pyridinedicarboxylic acid dimethyl ester, 2,6-pyridinedicarboxylic acid, dimethyl ester, dimethyl dipicolinate, dimethyl pyridine-2,6-carboxylate, chembl72405, dimethylpyridine-2,6-dicarboxylate, dipicolinic acid dimethyl ester
PubChem CID 79549
IUPAC Name dimethyl pyridine-2,6-dicarboxylate
SMILES COC(=O)C1=NC(=CC=C1)C(=O)OC

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 2,6-Pyridinedicarboxylate