missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl 2,5-Dioxahexanedioate 98.0+%, TCI America™
$167.76 - $480.92
Chemical Identifiers
| CAS | 88754-66-9 |
|---|---|
| Molecular Formula | C6H10O6 |
| Molecular Weight (g/mol) | 178.14 |
| MDL Number | MFCD00085447 |
| InChI Key | DOMLQXFMDFZAAL-UHFFFAOYSA-N |
| Synonym | 2,5-Dioxahexanedioic Acid Dimethyl Ester |
| PubChem CID | 263025 |
| IUPAC Name | 2-methoxycarbonyloxyethyl methyl carbonate |
| SMILES | COC(=O)OCCOC(=O)OC |
Chemical Identifiers
| 88754-66-9 | |
| 178.14 | |
| DOMLQXFMDFZAAL-UHFFFAOYSA-N | |
| 263025 | |
| COC(=O)OCCOC(=O)OC |
| C6H10O6 | |
| MFCD00085447 | |
| 2,5-Dioxahexanedioic Acid Dimethyl Ester | |
| 2-methoxycarbonyloxyethyl methyl carbonate |
Specifications
| 88754-66-9 | |
| 220°C | |
| MFCD00085447 | |
| 2,5-Dioxahexanedioic Acid Dimethyl Ester | |
| COC(=O)OCCOC(=O)OC | |
| 178.14 | |
| 178.14 | |
| Liquid |
| Colorless | |
| C6H10O6 | |
| 5 g | |
| DOMLQXFMDFZAAL-UHFFFAOYSA-N | |
| 2-methoxycarbonyloxyethyl methyl carbonate | |
| 263025 | |
| ≥98.0% (GC) | |
| Dimethyl 2,5-Dioxahexanedioate |
Safety and Handling
TSCA : No