missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl 2,3-Pyridinedicarboxylate 98.0+%, TCI America™
Supplier: TCI America P130325G
Specifications
| Dimethyl 2,3-Pyridinedicarboxylate | |
| 56°C | |
| MFCD00186549 | |
| 2,3-pyridinedicarboxylic acid dimethyl ester, dimethyl quinolinate, dimethyl 2,3-pyridinedicarboxylate, 2,3-bis methoxycarbonyl pyridine, 2,3-pyridinedicarboxylic acid, dimethyl ester, methylquinolinic acid, 2,3-dimethyl ester, 2,3-dimethyl pyridine-2,3-dicarboxylate, pyridine-2,3-dicarboxylic acid dimethyl ester, mqdme, 2,3-dimethyl methylquinolinate | |
| COC(=O)C1=CC=CN=C1C(=O)OC | |
| 195.17 | |
| 195.17 | |
| Crystalline Powder |
| 605-38-9 | |
| C9H9NO4 | |
| 25 g | |
| YLGIBCYHQZTFQL-UHFFFAOYSA-N | |
| 2,3-dimethyl pyridine-2,3-dicarboxylate | |
| 69058 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 605-38-9 | |
| 195.17 | |
| YLGIBCYHQZTFQL-UHFFFAOYSA-N | |
| 69058 | |
| COC(=O)C1=CC=CN=C1C(=O)OC |
| C9H9NO4 | |
| MFCD00186549 | |
| 2,3-pyridinedicarboxylic acid dimethyl ester, dimethyl quinolinate, dimethyl 2,3-pyridinedicarboxylate, 2,3-bis methoxycarbonyl pyridine, 2,3-pyridinedicarboxylic acid, dimethyl ester, methylquinolinic acid, 2,3-dimethyl ester, 2,3-dimethyl pyridine-2,3-dicarboxylate, pyridine-2,3-dicarboxylic acid dimethyl ester, mqdme, 2,3-dimethyl methylquinolinate | |
| 2,3-dimethyl pyridine-2,3-dicarboxylate |
Safety and Handling
TSCA : No