missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2,3-Pyridinedicarboxylate 98.0+%, TCI America™

Catalog No. p-7135066
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
P13035G 5 g
P130325G 25 g
2 options

Catalog No. P13035G

Supplier: TCI America P13035G

Only null left

Chemical Identifiers

CAS 605-38-9
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00186549
InChI Key YLGIBCYHQZTFQL-UHFFFAOYSA-N
Synonym 2,3-pyridinedicarboxylic acid dimethyl ester, dimethyl quinolinate, dimethyl 2,3-pyridinedicarboxylate, 2,3-bis methoxycarbonyl pyridine, 2,3-pyridinedicarboxylic acid, dimethyl ester, methylquinolinic acid, 2,3-dimethyl ester, 2,3-dimethyl pyridine-2,3-dicarboxylate, pyridine-2,3-dicarboxylic acid dimethyl ester, mqdme, 2,3-dimethyl methylquinolinate
PubChem CID 69058
IUPAC Name 2,3-dimethyl pyridine-2,3-dicarboxylate
SMILES COC(=O)C1=CC=CN=C1C(=O)OC

Specifications

Melting Point 56°C
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 2,3-Pyridinedicarboxylate