missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl (-)-2,3-O-Isopropylidene-L-tartrate 95.0+%, TCI America™
$96.94 - $369.08
Chemical Identifiers
| CAS | 37031-29-1 |
|---|---|
| Molecular Formula | C9H14O6 |
| Molecular Weight (g/mol) | 218.21 |
| MDL Number | MFCD00066218 |
| InChI Key | ROZOUYVVWUTPNG-PHDIDXHHSA-N |
| Synonym | 4r,5r-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, --dimethyl 2,3-o-isopropylidene-l-tartrate, dimethyl 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, --dimethyl-2,3-o-isopropylidene-l-tartrate, 4,5-dimethyl 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, pubchem6766, dimethyl--2,3-o-isopropylidene-l-tartrate, ksc917k0j, rozouyvvwutpng-phdidxhhsa |
| PubChem CID | 688155 |
| IUPAC Name | 4,5-dimethyl (4R,5R)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate |
| SMILES | COC(=O)[C@@H]1OC(C)(C)O[C@H]1C(=O)OC |
Chemical Identifiers
| 37031-29-1 | |
| 218.21 | |
| ROZOUYVVWUTPNG-PHDIDXHHSA-N | |
| 688155 | |
| COC(=O)[C@@H]1OC(C)(C)O[C@H]1C(=O)OC |
| C9H14O6 | |
| MFCD00066218 | |
| 4r,5r-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, --dimethyl 2,3-o-isopropylidene-l-tartrate, dimethyl 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, --dimethyl-2,3-o-isopropylidene-l-tartrate, 4,5-dimethyl 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, pubchem6766, dimethyl--2,3-o-isopropylidene-l-tartrate, ksc917k0j, rozouyvvwutpng-phdidxhhsa | |
| 4,5-dimethyl (4R,5R)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate |
Specifications
| 37031-29-1 | |
| 139°C | |
| MFCD00066218 | |
| 4r,5r-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, --dimethyl 2,3-o-isopropylidene-l-tartrate, dimethyl 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, --dimethyl-2,3-o-isopropylidene-l-tartrate, 4,5-dimethyl 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, pubchem6766, dimethyl--2,3-o-isopropylidene-l-tartrate, ksc917k0j, rozouyvvwutpng-phdidxhhsa | |
| COC(=O)[C@@H]1OC(C)(C)O[C@H]1C(=O)OC | |
| 218.21 | |
| 218.21 | |
| Liquid |
| Yellow | |
| C9H14O6 | |
| 5 g | |
| ROZOUYVVWUTPNG-PHDIDXHHSA-N | |
| 4,5-dimethyl (4R,5R)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate | |
| 688155 | |
| ≥95.0% (GC) | |
| Dimethyl (-)-2,3-O-Isopropylidene-L-tartrate |
Safety and Handling
TSCA : No