accessibility menu, dialog, popup

UserName

CAS RN 37031-29-1

H3COO(R)OH3CCH3O(R)OOCH3

CAS RN 37031-29-1

IUPAC Name: 4,5-dimethyl (4R,5R)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
Synonyms: 4,5-dimethyl (4R,5R)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
Molecular Weight (g/mol): 218.21
Molecular Formula: C9H14O6
InChi Key: ROZOUYVVWUTPNG-PHDIDXHHSA-N
SMILES: COC(=O)[C@@H]1OC(C)(C)O[C@H]1C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results