missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2,3-Naphthalenedicarboxylate 97.0+%, TCI America™

Catalog No. p-7133883
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05901G 1 g
N05905G 5 g
N059025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N05901G Supplier TCI America Supplier No. N05901G

Chemical Identifiers

CAS 13728-34-2
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD00012278
InChI Key MPDGBCOIHNLQMR-UHFFFAOYSA-N
Synonym 2,3-Naphthalenedicarboxylic Acid Dimethyl Ester
PubChem CID 688113
IUPAC Name dimethyl naphthalene-2,3-dicarboxylate
SMILES COC(=O)C1=CC2=CC=CC=C2C=C1C(=O)OC

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 145°C
Quantity 1 g
Formula Weight 244.25
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Dimethyl 2,3-Naphthalenedicarboxylate