missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl 2,2'-Bipyridine-4,4'-dicarboxylate 98.0+%, TCI America™
$188.27 - $624.46
Chemical Identifiers
| CAS | 71071-46-0 |
|---|---|
| Molecular Formula | C14H12N2O4 |
| Molecular Weight (g/mol) | 272.26 |
| MDL Number | MFCD02669916 |
| InChI Key | HBWBVIDKBKOVEX-UHFFFAOYSA-N |
| PubChem CID | 326419 |
| IUPAC Name | methyl 2-(4-methoxycarbonylpyridin-2-yl)pyridine-4-carboxylate |
| SMILES | COC(=O)C1=CC(=NC=C1)C2=NC=CC(=C2)C(=O)OC |
Chemical Identifiers
| 71071-46-0 | |
| 272.26 | |
| HBWBVIDKBKOVEX-UHFFFAOYSA-N | |
| methyl 2-(4-methoxycarbonylpyridin-2-yl)pyridine-4-carboxylate |
| C14H12N2O4 | |
| MFCD02669916 | |
| 326419 | |
| COC(=O)C1=CC(=NC=C1)C2=NC=CC(=C2)C(=O)OC |
Specifications
| 71071-46-0 | |
| White-Yellow | |
| MFCD02669916 | |
| 2,2′C-Bipyridine-4,4′C-dicarboxylic Acid Dimethyl Ester, 4,4′C-Bis(methoxycarbonyl)-2,2′C-bipyridine, 4,4′C-Dimethoxycarbonyl-2,2′C-bipyridine | |
| COC(=O)C1=CC(=NC=C1)C2=NC=CC(=C2)C(=O)OC | |
| 272.26 | |
| 272.26 | |
| Crystalline Powder |
| 210°C | |
| C14H12N2O4 | |
| 1 g | |
| HBWBVIDKBKOVEX-UHFFFAOYSA-N | |
| methyl 2-(4-methoxycarbonylpyridin-2-yl)pyridine-4-carboxylate | |
| 326419 | |
| ≥98.0% (T) | |
| Dimethyl 2,2′-Bipyridine-4,4′-dicarboxylate |
Safety and Handling
TSCA : No