accessibility menu, dialog, popup

UserName

CAS RN 71071-46-0

CH3OONNOOCH3

CAS RN 71071-46-0

IUPAC Name: 4,4'-dimethyl [2,2'-bipyridine]-4,4'-dicarboxylate
Synonyms: 4,4'-dimethyl [2,2'-bipyridine]-4,4'-dicarboxylate
Molecular Weight (g/mol): 272.26
Molecular Formula: C14H12N2O4
InChi Key: HBWBVIDKBKOVEX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results