missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2,2'-Bipyridine-4,4'-dicarboxylate 98.0+%, TCI America™

Catalog No. D46355G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46355G 5 g
D46351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46355G Supplier TCI America Supplier No. D46355G
Only null left

Chemical Identifiers

CAS 71071-46-0
Molecular Formula C14H12N2O4
Molecular Weight (g/mol) 272.26
MDL Number MFCD02669916
InChI Key HBWBVIDKBKOVEX-UHFFFAOYSA-N
PubChem CID 326419
IUPAC Name methyl 2-(4-methoxycarbonylpyridin-2-yl)pyridine-4-carboxylate
SMILES COC(=O)C1=CC(=NC=C1)C2=NC=CC(=C2)C(=O)OC

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 5 g
Synonym 2,2′-Bipyridine-4,4′-dicarboxylic Acid Dimethyl Ester, 4,4′-Bis(methoxycarbonyl)-2,2′-bipyridine, 4,4′-Dimethoxycarbonyl-2,2′-bipyridine
Formula Weight 272.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 2,2′-Bipyridine-4,4′-dicarboxylate