Learn More
Dimethyl (1-diazo-2-oxopropyl)phosphonate, 95%, Thermo Scientific™
Chemical Identifiers
| CAS | 90965-06-3 |
|---|---|
| Molecular Formula | C5H9N2O4P |
| Molecular Weight (g/mol) | 192.11 |
| InChI Key | SQHSJJGGWYIFCD-UHFFFAOYSA-N |
| IUPAC Name | 1-(diazyn-1-ium-1-yl)-1-(dimethoxyphosphoryl)prop-1-en-2-olate |
| SMILES | COP(=O)(OC)C([N+]#N)=C(C)[O-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC466020010
|
N/A | 1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC466020050
|
N/A | 5 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 90965-06-3 | |
| 192.11 | |
| 1-(diazyn-1-ium-1-yl)-1-(dimethoxyphosphoryl)prop-1-en-2-olate |
| C5H9N2O4P | |
| SQHSJJGGWYIFCD-UHFFFAOYSA-N | |
| COP(=O)(OC)C([N+]#N)=C(C)[O-] |
Specifications
| 90965-06-3 | |
| C5H9N2O4P | |
| COP(=O)(OC)C([N+]#N)=C(C)[O-] | |
| 192.11 | |
| Dimethyl (1-diazo-2-oxopropyl)phosphonate |
| 95% | |
| SQHSJJGGWYIFCD-UHFFFAOYSA-N | |
| 1-(diazyn-1-ium-1-yl)-1-(dimethoxyphosphoryl)prop-1-en-2-olate | |
| 192.11 |
Safety and Handling
GHS H Statement Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
RUO – Research Use Only