missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (1-diazo-2-oxopropyl)phosphonate, 95%

Catalog No. AC466020010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466020010 1 g
AC466020050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466020010 Supplier Thermo Scientific Chemicals Supplier No. 466020010

CAS: 90965-06-3 | C5H9N2O4P | 192.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90965-06-3
Molecular Formula C5H9N2O4P
Molecular Weight (g/mol) 192.11
InChI Key SQHSJJGGWYIFCD-UHFFFAOYSA-N
IUPAC Name 1-(diazyn-1-ium-1-yl)-1-(dimethoxyphosphoryl)prop-1-en-2-olate
SMILES COP(=O)(OC)C([N+]#N)=C(C)[O-]

Specifications

Assay Percent Range 95%
Quantity 1 g
Formula Weight 192.11
Chemical Name or Material Dimethyl (1-diazo-2-oxopropyl)phosphonate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Harmful in contact with skin.

Harmful if inhaled.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only