missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,8-Anthracenedicarboxylate 95.0+%, TCI America™

Catalog No. p-7117433
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14331G 1 g
A14335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14331G Supplier TCI America Supplier No. A14331G
Only null left

Chemical Identifiers

CAS 93655-34-6
Molecular Formula C18H14O4
Molecular Weight (g/mol) 294.306
MDL Number MFCD01321126
InChI Key ALYSIEHCBBJETD-UHFFFAOYSA-N
Synonym 1,8-Anthracenedicarboxylic Acid Dimethyl Ester
PubChem CID 5260121
IUPAC Name dimethyl anthracene-1,8-dicarboxylate
SMILES COC(=O)C1=CC=CC2=CC3=C(C=C21)C(=CC=C3)C(=O)OC

Specifications

Melting Point 105°C
Color Yellow
Quantity 1 g
Formula Weight 294.31
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 1,8-Anthracenedicarboxylate