accessibility menu, dialog, popup

UserName

CAS RN 93655-34-6

IUPAC Name: 1,8-dimethyl anthracene-1,8-dicarboxylate
Synonyms: 1,8-dimethyl anthracene-1,8-dicarboxylate
Molecular Weight (g/mol): 294.31
Molecular Formula: C18H14O4
InChi Key: ALYSIEHCBBJETD-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC2=CC3=CC=CC(C(=O)OC)=C3C=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results