missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,5-Naphthalenedisulfonate 98.0+%, TCI America™

Catalog No. N043925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N043925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N043925G Supplier TCI America Supplier No. N043925G
Only null left

Chemical Identifiers

CAS 20779-13-9
Molecular Formula C12H12O6S2
Molecular Weight (g/mol) 316.342
MDL Number MFCD00059544
InChI Key HCXPJMZQMWIBMO-UHFFFAOYSA-N
Synonym 1,5-Naphthalenedisulfonic Acid Dimethyl Ester
PubChem CID 245329
IUPAC Name dimethyl naphthalene-1,5-disulfonate
SMILES COS(=O)(=O)C1=CC=CC2=C1C=CC=C2S(=O)(=O)OC

Specifications

Melting Point 203°C
Color White
Quantity 25 g
Formula Weight 316.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 1,5-Naphthalenedisulfonate