missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,4-Naphthalenedicarboxylate 98.0+%, TCI America™

Catalog No. p-7128256
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44001G 1 g
D44005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44001G Supplier TCI America Supplier No. D44001G
Only null left

Chemical Identifiers

CAS 7487-15-2
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD04038075
InChI Key OCSXMIBZIHMVCP-UHFFFAOYSA-N
Synonym 1,4-Naphthalenedicarboxylic Acid Dimethyl Ester
PubChem CID 11064621
IUPAC Name dimethyl naphthalene-1,4-dicarboxylate
SMILES COC(=O)C1=CC=C(C2=CC=CC=C21)C(=O)OC

Specifications

Melting Point 67°C
Color White-Yellow
Boiling Point 161°C
Quantity 1 g
Formula Weight 244.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 1,4-Naphthalenedicarboxylate