missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,3-Dithiole-2-thione-4,5-dicarboxylate 98.0+%, TCI America™

Catalog No. D212725G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D212725G 25 g
D212710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D212725G Supplier TCI America Supplier No. D212725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7396-41-0
Molecular Formula C7H6O4S3
Molecular Weight (g/mol) 250.301
MDL Number MFCD00142532
InChI Key UZKKFHMMXWDIPD-UHFFFAOYSA-N
Synonym Dimethyl 2-Thioxo-1,3-dithiole-4,5-dicarboxylate, 1,3-Dithiole-2-thione-4,5-dicarboxylic Acid Dimethyl Ester
PubChem CID 547914
IUPAC Name dimethyl 2-sulfanylidene-1,3-dithiole-4,5-dicarboxylate
SMILES COC(=O)C1=C(SC(=S)S1)C(=O)OC

Specifications

Melting Point 88°C
Quantity 25 g
Formula Weight 250.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 1,3-Dithiole-2-thione-4,5-dicarboxylate