accessibility menu, dialog, popup

UserName

CAS RN 7396-41-0

IUPAC Name: 4,5-dimethyl 2-sulfanylidene-2H-1,3-dithiole-4,5-dicarboxylate
Synonyms: 4,5-dimethyl 2-sulfanylidene-1,3-dithiole-4,5-dicarboxylate
Molecular Weight (g/mol): 250.3
Molecular Formula: C7H6O4S3
InChi Key: UZKKFHMMXWDIPD-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(SC(=S)S1)C(=O)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results