missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,3-Acetonedicarboxylate 95.0+%, TCI America™

Catalog No. p-7116685
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A005625G 25 g
A0056100G 100 g
A0056500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A005625G Supplier TCI America Supplier No. A005625G
Only null left

Chemical Identifiers

CAS 1830-54-2
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.15
MDL Number MFCD00008462
InChI Key RNJOKCPFLQMDEC-UHFFFAOYSA-N
Synonym dimethyl 1,3-acetonedicarboxylate, dimethyl acetone-1,3-dicarboxylate, dimethyl 3-oxoglutarate, 1,5-dimethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, dimethyl ester, dimethyl acetonedicarboxylate, acetone dicarboxylic acid, dimethyl ester, glutaric acid, 3-oxo-, dimethyl ester, pentanedioic acid, 3-oxo-, 1,5-dimethyl ester, dimethyl 3-oxopentane-1,5-dioate
PubChem CID 74591
IUPAC Name 1,5-dimethyl 3-oxopentanedioate
SMILES COC(=O)CC(=O)CC(=O)OC

Specifications

Color Yellow
Boiling Point 150°C
Quantity 25 g
Formula Weight 174.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl 1,3-Acetonedicarboxylate
EINECSNumber (2)-3286
TSCA Yes
Recommended Storage Refrigerator