missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,3-Acetonedicarboxylate 95.0+%, TCI America™

Supplier:  TCI America A0056100G

Quantity 100 g

 View more versions of this product

Catalog No. A0056100G


Only null left

Description

Description

Specifications

Chemical Identifiers

1830-54-2
174.15
RNJOKCPFLQMDEC-UHFFFAOYSA-N
74591
COC(=O)CC(=O)CC(=O)OC
C7H10O5
MFCD00008462
dimethyl 1,3-acetonedicarboxylate, dimethyl acetone-1,3-dicarboxylate, dimethyl 3-oxoglutarate, 1,5-dimethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, dimethyl ester, dimethyl acetonedicarboxylate, acetone dicarboxylic acid, dimethyl ester, glutaric acid, 3-oxo-, dimethyl ester, pentanedioic acid, 3-oxo-, 1,5-dimethyl ester, dimethyl 3-oxopentane-1,5-dioate
1,5-dimethyl 3-oxopentanedioate

Specifications

Dimethyl 1,3-Acetonedicarboxylate
Yellow
C7H10O5
MFCD00008462
RNJOKCPFLQMDEC-UHFFFAOYSA-N
1,5-dimethyl 3-oxopentanedioate
74591
≥95.0% (GC)
1830-54-2
150°C
100 g
dimethyl 1,3-acetonedicarboxylate, dimethyl acetone-1,3-dicarboxylate, dimethyl 3-oxoglutarate, 1,5-dimethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, dimethyl ester, dimethyl acetonedicarboxylate, acetone dicarboxylic acid, dimethyl ester, glutaric acid, 3-oxo-, dimethyl ester, pentanedioic acid, 3-oxo-, 1,5-dimethyl ester, dimethyl 3-oxopentane-1,5-dioate
COC(=O)CC(=O)CC(=O)OC
174.15
174.15
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3286

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions