missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxydimethylsilane 98.0+%, TCI America™

Catalog No. D1052500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1052500ML 500 mL
D105225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1052500ML Supplier TCI America Supplier No. D1052500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1112-39-6
Molecular Formula C4H12O2Si
Molecular Weight (g/mol) 120.22
MDL Number MFCD00025691
InChI Key JJQZDUKDJDQPMQ-UHFFFAOYSA-N
Synonym dimethyldimethoxysilane, silane, dimethoxydimethyl, kbm 22, unii-a3qdb1rs1c, dimethyl dimethoxysilane, dimethoxy dimethyl silane, a3qdb1rs1c, dimethoxy-dimethylsilane, dimethyl dimethoxy silane, dimethyl dimethoxy silicane
PubChem CID 66187
IUPAC Name dimethoxydimethylsilane
SMILES CO[Si](C)(C)OC

Specifications

Color Colorless
Boiling Point 81°C
Quantity 500 mL
UN Number 1993
Formula Weight 120.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethoxydimethylsilane
EINECSNumber (2)-2053&(2)-2052
TSCA Yes