accessibility menu, dialog, popup

UserName

CH3OSiCH3H3COH3C

CAS RN 1112-39-6

IUPAC Name: dimethoxydimethylsilane
Synonyms: silane, dimethoxydimethyl-
Molecular Weight (g/mol): 120.22
Molecular Formula: C4H12O2Si
InChi Key: JJQZDUKDJDQPMQ-UHFFFAOYSA-N
SMILES: CO[Si](C)(C)OC
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)