missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxydi-p-tolylsilane 98.0+%, TCI America™

Catalog No. p-7127885
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37331G 1 g
D37335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37331G Supplier TCI America Supplier No. D37331G

Chemical Identifiers

CAS 92779-72-1
Molecular Formula C16H20O2Si
Molecular Weight (g/mol) 272.419
MDL Number MFCD01861714
InChI Key SZIZIGBTHTUEBU-UHFFFAOYSA-N
PubChem CID 11821755
IUPAC Name dimethoxy-bis(4-methylphenyl)silane
SMILES CC1=CC=C(C=C1)[Si](C2=CC=C(C=C2)C)(OC)OC

Specifications

Color Colorless
Quantity 1 g
Formula Weight 272.42
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethoxydi-p-tolylsilane