accessibility menu, dialog, popup

UserName

CAS RN 92779-72-1

CH3SiCH3OH3COCH3

CAS RN 92779-72-1

IUPAC Name: dimethoxybis(4-methylphenyl)silane
Synonyms: dimethoxybis(4-methylphenyl)silane
Molecular Weight (g/mol): 272.42
Molecular Formula: C16H20O2Si
InChi Key: SZIZIGBTHTUEBU-UHFFFAOYSA-N
SMILES: CO[Si](OC)(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results