missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethoxy(methyl)silane 97.0+%, TCI America™
$272.16 - $928.30
Chemical Identifiers
| CAS | 16881-77-9 |
|---|---|
| Molecular Formula | C3H9O2Si |
| Molecular Weight (g/mol) | 105.188 |
| MDL Number | MFCD00048019 |
| InChI Key | PKTOVQRKCNPVKY-UHFFFAOYSA-N |
| Synonym | methyldimethoxysilane, dimethoxy methyl silane, silane, dimethoxymethyl, dimethoxy methyl silicon, methyl dimethoxysilane, c3h10o2si, methyl dimethoxy silane, dimethoxy methyl silane #, dimethoxy methyl silyl |
| PubChem CID | 6329250 |
| IUPAC Name | dimethoxy(methyl)silicon |
| SMILES | CO[Si](C)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D210025ML
|
TCI America
D210025ML |
25 mL |
Each for $272.16
|
|
|||||
|
D2100100ML
|
TCI America
D2100100ML |
100 mL |
Each for $928.30
|
|
|||||
Chemical Identifiers
| 16881-77-9 | |
| 105.188 | |
| PKTOVQRKCNPVKY-UHFFFAOYSA-N | |
| 6329250 | |
| CO[Si](C)OC |
| C3H9O2Si | |
| MFCD00048019 | |
| methyldimethoxysilane, dimethoxy methyl silane, silane, dimethoxymethyl, dimethoxy methyl silicon, methyl dimethoxysilane, c3h10o2si, methyl dimethoxy silane, dimethoxy methyl silane #, dimethoxy methyl silyl | |
| dimethoxy(methyl)silicon |
Specifications
| 16881-77-9 | |
| 61°C | |
| MFCD00048019 | |
| 1992 | |
| PKTOVQRKCNPVKY-UHFFFAOYSA-N | |
| dimethoxy(methyl)silicon | |
| 6329250 | |
| ≥97.0% (GC) | |
| Dimethoxy(methyl)silane |
| Colorless | |
| C3H9O2Si | |
| 25 mL | |
| methyldimethoxysilane, dimethoxy methyl silane, silane, dimethoxymethyl, dimethoxy methyl silicon, methyl dimethoxysilane, c3h10o2si, methyl dimethoxy silane, dimethoxy methyl silane #, dimethoxy methyl silyl | |
| CO[Si](C)OC | |
| 105.188 | |
| 106.20 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2052&(2)-2053
RTECSNumber : VV3644166
TSCA : Yes