missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethoxy(methyl)silane 97.0+%, TCI America™
Supplier: TCI America D2100100ML
Specifications
| Dimethoxy(methyl)silane | |
| Colorless | |
| C3H9O2Si | |
| 100 mL | |
| methyldimethoxysilane, dimethoxy methyl silane, silane, dimethoxymethyl, dimethoxy methyl silicon, methyl dimethoxysilane, c3h10o2si, methyl dimethoxy silane, dimethoxy methyl silane #, dimethoxy methyl silyl | |
| CO[Si](C)OC | |
| 105.188 | |
| 106.20 | |
| Liquid |
| 16881-77-9 | |
| 61°C | |
| MFCD00048019 | |
| 1992 | |
| PKTOVQRKCNPVKY-UHFFFAOYSA-N | |
| dimethoxy(methyl)silicon | |
| 6329250 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 16881-77-9 | |
| 105.188 | |
| PKTOVQRKCNPVKY-UHFFFAOYSA-N | |
| 6329250 | |
| CO[Si](C)OC |
| C3H9O2Si | |
| MFCD00048019 | |
| methyldimethoxysilane, dimethoxy methyl silane, silane, dimethoxymethyl, dimethoxy methyl silicon, methyl dimethoxysilane, c3h10o2si, methyl dimethoxy silane, dimethoxy methyl silane #, dimethoxy methyl silyl | |
| dimethoxy(methyl)silicon |
Safety and Handling
EINECSNumber : (2)-2052&(2)-2053
RTECSNumber : VV3644166
TSCA : Yes