missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethoxy(methyl)(3,3,3-trifluoropropyl)silane 98.0+%, TCI America™
Supplier: TCI America D532825G
Specifications
| Dimethoxy(methyl)(3,3,3-trifluoropropyl)silane | |
| Colorless | |
| C6H13F3O2Si | |
| 25 g | |
| dimethoxy methyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl methyldimethoxysilane, 3,3,3-trifluoropropylmethyldimethoxysilane, dimethoxymethyl 3,3,3-trifluoropropyl silane, silane, dimethoxymethyl 3,3,3-trifluoropropyl, dimethoxy-methyl-3,3,3-trifluoropropyl silane, acmc-1cm60, 3,3,3-trifluoropropyldimethoxy methyl silane, methyl 3,3,3-trifluoropropyl dimethoxysilane, silane,dimethoxymethyl 3,3,3-trifluoropropyl | |
| CO[Si](C)(CCC(F)(F)F)OC | |
| 202.248 | |
| 202.25 | |
| Liquid |
| 358-67-8 | |
| 85°C | |
| MFCD00053175 | |
| 1993 | |
| DIJRHOZMLZRNLM-UHFFFAOYSA-N | |
| dimethoxy-methyl-(3,3,3-trifluoropropyl)silane | |
| 67751 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 358-67-8 | |
| 202.248 | |
| DIJRHOZMLZRNLM-UHFFFAOYSA-N | |
| 67751 | |
| CO[Si](C)(CCC(F)(F)F)OC |
| C6H13F3O2Si | |
| MFCD00053175 | |
| dimethoxy methyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl methyldimethoxysilane, 3,3,3-trifluoropropylmethyldimethoxysilane, dimethoxymethyl 3,3,3-trifluoropropyl silane, silane, dimethoxymethyl 3,3,3-trifluoropropyl, dimethoxy-methyl-3,3,3-trifluoropropyl silane, acmc-1cm60, 3,3,3-trifluoropropyldimethoxy methyl silane, methyl 3,3,3-trifluoropropyl dimethoxysilane, silane,dimethoxymethyl 3,3,3-trifluoropropyl | |
| dimethoxy-methyl-(3,3,3-trifluoropropyl)silane |
Safety and Handling
EINECSNumber : (2)-2078
TSCA : Yes