missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxy(methyl)(3,3,3-trifluoropropyl)silane 98.0+%, TCI America™

Catalog No. p-7128733
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53285G 5 g
D532825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53285G Supplier TCI America Supplier No. D53285G

Chemical Identifiers

CAS 358-67-8
Molecular Formula C6H13F3O2Si
Molecular Weight (g/mol) 202.248
MDL Number MFCD00053175
InChI Key DIJRHOZMLZRNLM-UHFFFAOYSA-N
Synonym dimethoxy methyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl methyldimethoxysilane, 3,3,3-trifluoropropylmethyldimethoxysilane, dimethoxymethyl 3,3,3-trifluoropropyl silane, silane, dimethoxymethyl 3,3,3-trifluoropropyl, dimethoxy-methyl-3,3,3-trifluoropropyl silane, acmc-1cm60, 3,3,3-trifluoropropyldimethoxy methyl silane, methyl 3,3,3-trifluoropropyl dimethoxysilane, silane,dimethoxymethyl 3,3,3-trifluoropropyl
PubChem CID 67751
IUPAC Name dimethoxy-methyl-(3,3,3-trifluoropropyl)silane
SMILES CO[Si](C)(CCC(F)(F)F)OC

Specifications

Color Colorless
Boiling Point 85°C
Quantity 5 g
UN Number 1993
Formula Weight 202.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethoxy(methyl)(3,3,3-trifluoropropyl)silane
EINECSNumber (2)-2078
TSCA Yes