missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimesitylammonium Pentafluorobenzenesulfonate 98.0+%, TCI America™
$281.48 - $1015.90
Chemical Identifiers
| CAS | 850629-65-1 |
|---|---|
| Molecular Formula | C24H24F5NO3S |
| Molecular Weight (g/mol) | 501.512 |
| InChI Key | MKKHTKBUMNIBLN-UHFFFAOYSA-N |
| PubChem CID | 44629836 |
| IUPAC Name | bis(2,4,6-trimethylphenyl)azanium;2,3,4,5,6-pentafluorobenzenesulfonate |
| SMILES | CC1=CC(=C(C(=C1)C)[NH2+]C2=C(C=C(C=C2C)C)C)C.C1(=C(C(=C(C(=C1F)F)S(=O)(=O)[O-])F)F)F |
Chemical Identifiers
| 850629-65-1 | |
| 501.512 | |
| 44629836 | |
| CC1=CC(=C(C(=C1)C)[NH2+]C2=C(C=C(C=C2C)C)C)C.C1(=C(C(=C(C(=C1F)F)S(=O)(=O)[O-])F)F)F |
| C24H24F5NO3S | |
| MKKHTKBUMNIBLN-UHFFFAOYSA-N | |
| bis(2,4,6-trimethylphenyl)azanium;2,3,4,5,6-pentafluorobenzenesulfonate |
Specifications
| 850629-65-1 | |
| 1 g | |
| CC1=CC(=C(C(=C1)C)[NH2+]C2=C(C=C(C=C2C)C)C)C.C1(=C(C(=C(C(=C1F)F)S(=O)(=O)[O-])F)F)F | |
| 501.512 | |
| 501.51 | |
| Crystalline Powder |
| C24H24F5NO3S | |
| MKKHTKBUMNIBLN-UHFFFAOYSA-N | |
| bis(2,4,6-trimethylphenyl)azanium;2,3,4,5,6-pentafluorobenzenesulfonate | |
| 44629836 | |
| ≥98.0% (T) | |
| Dimesitylammonium Pentafluorobenzenesulfonate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator