accessibility menu, dialog, popup

UserName

CAS RN 850629-65-1

CH3H3CNH2H3CH3CCH3CH3FFSOOOFFF

CAS RN 850629-65-1

IUPAC Name: 2,4,6-trimethyl-N-(2,4,6-trimethylphenyl)anilinium pentafluorobenzene-1-sulfonate
Synonyms: 2,4,6-trimethyl-N-(2,4,6-trimethylphenyl)anilinium pentafluorobenzenesulfonate
Molecular Weight (g/mol): 501.51
Molecular Formula: C24H24F5NO3S
InChi Key: MKKHTKBUMNIBLN-UHFFFAOYSA-N
SMILES: [O-]S(=O)(=O)C1=C(F)C(F)=C(F)C(F)=C1F.CC1=CC(C)=C([NH2+]C2=C(C)C=C(C)C=C2C)C(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results