missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Dimedone | |
| 149°C | |
| C8H12O2 | |
| 100 g | |
| BADXJIPKFRBFOT-UHFFFAOYSA-N | |
| 5,5-dimethylcyclohexane-1,3-dione | |
| 31358 | |
| ≥99.0% (T) |
| 126-81-8 | |
| White-Yellow | |
| MFCD00001588 | |
| dimedone, 5,5-dimethyl-1,3-cyclohexanedione, cyclomethone, medon, methone, dimedon, methon, 1,3-cyclohexanedione, 5,5-dimethyl, 5,5-dimethyldihydroresorcinol, 5,5-dimethylhydroresorcinol | |
| CC1(C)CC(=O)CC(=O)C1 | |
| 140.18 | |
| 140.18 | |
| Crystalline Powder |
Chemical Identifiers
| 126-81-8 | |
| 140.18 | |
| BADXJIPKFRBFOT-UHFFFAOYSA-N | |
| 31358 | |
| CC1(C)CC(=O)CC(=O)C1 |
| C8H12O2 | |
| MFCD00001588 | |
| dimedone, 5,5-dimethyl-1,3-cyclohexanedione, cyclomethone, medon, methone, dimedon, methon, 1,3-cyclohexanedione, 5,5-dimethyl, 5,5-dimethyldihydroresorcinol, 5,5-dimethylhydroresorcinol | |
| 5,5-dimethylcyclohexane-1,3-dione |
Safety and Handling
EINECSNumber : (3)-2374
RTECSNumber : GV0345000
TSCA : Yes