missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diluted Hydrochloric Acid, 10% (w/v), NF, 9.5-150%, Spectrum™ Chemical
HY105, 7647-01-0, HCl
Supplier: Spectrum Chemical Mfg Cor HY10525LTPVCS4
Description
Spectrum™ Chemical Diluted Hydrochloric Acid, 10 Percent (w/v), NF is a pH acidifier excipient in pharmaceutical preparations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
| Hydrochloric Acid | |
| 10%,90% | |
| 4 x 2.5 L | |
| MFCD00011324 MFCD00792839 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 9.5 to 10.5% |
| 7647-01-0,7732-18-5 | |
| Poly-Coated Glass Bottle | |
| ClH | |
| UN1789 | |
| Cl | |
| 36.46 | |
| NF |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| Cl |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrogen chloride |