Learn More
Thermo Scientific Chemicals Diltiazem hydrochloride, 98%
CAS: 33286-22-5 | C22H26N2O4S·HCl | 450.99 g/mol
Supplier: Thermo Scientific Chemicals 329080010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Recyclable
Specifications
Diltiazem hydrochloride | |
210.0°C to 215.0°C | |
0.1% max. | |
Authentic | |
Glass bottle | |
MFCD00069252 | |
15,3224 | |
Solubility in water: soluble. Other solubilities: soluble in chloroform,formic acid,methanol | |
CC(=O)OC1C(SC2=CC=CC=C2N(C1=O)CCN(C)C)C3=CC=C(C=C3)OC.Cl | |
450.99 | |
CHEBI:645509 | |
98% |
33286-22-5 | |
White | |
0.5% max. | |
97.5% min. (HPLC) | |
C22H26N2O4S·HCl | |
1 g | |
diltiazem hydrochloride, diltiazem hcl, herbesser, dilzene, lacerol, masdil, tildiem, tiazac, mono-tildiem, bi-tildiem | |
HDRXZJPWHTXQRI-BHDTVMLSSA-N | |
[(2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate;hydrochloride | |
62920 | |
450.99 | |
Powder |
Chemical Identifiers
33286-22-5 | |
450.99 | |
HDRXZJPWHTXQRI-BHDTVMLSSA-N | |
62920 | |
[(2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate;hydrochlorideShow More |
C22H26N2O4S·HCl | |
MFCD00069252 | |
diltiazem hydrochloride, diltiazem hcl, herbesser, dilzene, lacerol, masdil, tildiem, tiazac, mono-tildiem, bi-tildiemShow More | |
CHEBI:645509 | |
CC(=O)OC1C(SC2=CC=CC=C2N(C1=O)CCN(C)C)C3=CC=C(C=C3)OC.Cl |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 251-443-3
RUO – Research Use Only