missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Diloxanide (furoate), 97%
$338.95 - $1006.22
Chemical Identifiers
| CAS | 3736-81-0 |
|---|---|
| Molecular Formula | C14H11Cl2NO4 |
| Molecular Weight (g/mol) | 328.15 |
| MDL Number | MFCD00083302 |
| InChI Key | BDYYDXJSHYEDGB-UHFFFAOYSA-N |
| IUPAC Name | 4-(2,2-dichloro-N-methylacetamido)phenyl furan-2-carboxylate |
| SMILES | CN(C(=O)C(Cl)Cl)C1=CC=C(OC(=O)C2=CC=CO2)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH36096MD
|
Thermo Scientific™
H36096MD |
250 mg |
Each for $338.95
|
|
|||||
|
AAH3609603
|
Thermo Scientific™
H3609603 |
1 g |
Each for $1,006.22
|
|
|||||
Chemical Identifiers
| 3736-81-0 | |
| 328.15 | |
| BDYYDXJSHYEDGB-UHFFFAOYSA-N | |
| CN(C(=O)C(Cl)Cl)C1=CC=C(OC(=O)C2=CC=CO2)C=C1 |
| C14H11Cl2NO4 | |
| MFCD00083302 | |
| 4-(2,2-dichloro-N-methylacetamido)phenyl furan-2-carboxylate |
Specifications
| 3736-81-0 | |
| MFCD00083302 | |
| BDYYDXJSHYEDGB-UHFFFAOYSA-N | |
| 4-(2,2-dichloro-N-methylacetamido)phenyl furan-2-carboxylate | |
| 328.15 | |
| Solid |
| C14H11Cl2NO4 | |
| 250 mg | |
| CN(C(=O)C(Cl)Cl)C1=CC=C(OC(=O)C2=CC=CO2)C=C1 | |
| 328.15 | |
| 97% |
Safety and Handling
EINECSNumber : 223-108-1