missing translation for 'onlineSavingsMsg'
Learn More

Dilithium tetrachlorocuprate, 0.1M solution in THF

Catalog No. AC407830250
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC407830250 25 mL
AC407831000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC407830250 Supplier Thermo Scientific Chemicals Supplier No. 407830250

CAS: 15489-27-7 | Cl4CuLi2 | 219.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Dilithium tetrachlorocuprate
Color Colorless
Name Note 0.1M Solution in THF
CAS 15489-27-7, 109-99-9
Molecular Formula Cl4CuLi2
Linear Formula Li2CuCl4
MDL Number MFCD00011081
Concentration 0.09 to 0.12M
Fieser 04,163; 05,226; 06,203; 07,114; 08,176; 11,190; 12,195
Density 0.9100g/mL
InChI Key HCJWWBBBSCXJMS-UHFFFAOYSA-J
SMILES [Li+].[Li+].Cl[Cu-2](Cl)(Cl)Cl
IUPAC Name dilithium;tetrachlorocopper(2-)
Molecular Weight (g/mol) 219.226
PubChem CID 11074879
Formula Weight 219.24
Packaging Glass bottle
Specific Gravity 0.91
Quantity 25 mL
Flash Point −17°C
Physical Form Liquid
Show More Show Less

RUO – Research Use Only