missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyldichlorosilane, 97%

Catalog No. AC432360050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432360050 5 g Glass bottle
AC432360250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432360050 Supplier Thermo Scientific Chemicals Supplier No. 432360050

CAS: 7751-38-4 | C6H14Cl2Si | 185.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7751-38-4
Molecular Formula C6H14Cl2Si
Molecular Weight (g/mol) 185.16
MDL Number MFCD00054895
InChI Key GSENNYNYEKCQGA-UHFFFAOYSA-N
Synonym diisopropyldichlorosilane, dichlorodiisopropylsilane, silane, dichlorobis 1-methylethyl, dichloro-di propan-2-yl silane, silane, dichlorodiisopropyl, acmc-209pad, dichloro diisopropyl silane, di 2-propyl dichlorosilane, dichlorobis propan-2-yl silane, bis chloranyl-di propan-2-yl silane
PubChem CID 2758384
SMILES CC(C)[Si](Cl)(Cl)C(C)C

Specifications

Color Colorless
Density 1.0260g/mL
Boiling Point 168.0°C to 170.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4430 to 1.4450 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.026
IUPAC Name dichlorobis(propan-2-yl)silane
Formula Weight 185.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diisopropyldichlorosilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.

RUO – Research Use Only